Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTNNA1
Entrez Gene ID
100328952
Gene Name
catenin alpha 1
Gene Aliases
-
Chromosome Location
Chr.3: 21032448 - 21172627 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171142.1 NP_001164613.1 5 - 6 867 73
XM_008254892.2 XP_008253114.1 5 - 6 876 73

View Full Product Details