Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP2C
Entrez Gene ID
100328933
Gene Name
cytochrome P450-like
Gene Aliases
CYP2C3, CYPIIC3
Chromosome Location
Chr.18: 43496399 - 43525090 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171265.1 NP_001164736.1 5 - 6 848 96
GenBank mRNA D26152.1 - 5 - 6 848 96
K01523.1 - 4 - 5 561 96

View Full Product Details