Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STEAP1
Entrez Gene ID
100328630
Gene Name
STEAP family member 1
Gene Aliases
-
Chromosome Location
Chr.10: 38452824 - 38461647 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171274.1 NP_001164745.1 2 - 3 596 74
XM_017344299.1 XP_017199788.1 3 - 4 726 74

View Full Product Details