Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYSMD1
Entrez Gene ID
100328664
Gene Name
LysM domain containing 1
Gene Aliases
-
Chromosome Location
Chr.13: 41027024 - 41039753 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171297.1 NP_001164768.1 1 - 2 178 61
XM_008264017.2 XP_008262239.1 2 - 3 601 61
XM_008264018.2 XP_008262240.1 2 - 3 608 61

View Full Product Details