Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YWHAH
Entrez Gene ID
100328734
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein eta
Gene Aliases
-
Chromosome Location
Chr.21: 4316205 - 4325994 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171356.1 NP_001164827.1 1 - 2 84 85

View Full Product Details