Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRDM4
Entrez Gene ID
100328737
Gene Name
PR/SET domain 4
Gene Aliases
-
Chromosome Location
Chr.4: 88759098 - 88791402 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171358.1 NP_001164829.1 7 - 8 1621 86
XM_008257056.2 XP_008255278.1 8 - 9 1686 86
XM_008257057.2 XP_008255279.1 9 - 10 1923 86
XM_008257059.2 XP_008255281.1 7 - 8 1540 86
XM_017341624.1 XP_017197113.1 8 - 9 1946 86
XM_017341625.1 XP_017197114.1 8 - 9 1692 86

View Full Product Details