Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAZ
Entrez Gene ID
100328759
Gene Name
tafazzin
Gene Aliases
-
Chromosome Location
Chr.Unknown: 134225 - 141546 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171376.1 NP_001164847.1 - 101 56
XM_017338558.1 XP_017194047.1 - 455 56
XM_017338559.1 XP_017194048.1 - 455 56
XM_017338560.1 XP_017194049.1 - 455 56
XM_017338562.1 XP_017194051.1 - 453 56

View Full Product Details