Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB39B
Entrez Gene ID
100328771
Gene Name
RAB39B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.Unknown: 545276 - 548557 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171387.1 NP_001164858.1 - 209 92

View Full Product Details