Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNG7
Entrez Gene ID
100328774
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 7
Gene Aliases
-
Chromosome Location
Chr.Unknown: 119016 - 136300 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171389.1 NP_001164860.1 - 194 94

View Full Product Details