Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MDFIC
Entrez Gene ID
100328794
Gene Name
MyoD family inhibitor domain containing
Gene Aliases
-
Chromosome Location
Chr.7: 30351458 - 30452557 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171407.1 NP_001164878.1 2 - 3 215 74
XM_008258188.2 XP_008256410.1 3 - 4 759 74

View Full Product Details