Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR51S1
Entrez Gene ID
100328829
Gene Name
olfactory receptor, family 51, subfamily S, member 1
Gene Aliases
-
Chromosome Location
Chr.1: 145659192 - 145660162 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171431.1 NP_001164902.1 1 - 1 851 86

View Full Product Details