Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR52J3
Entrez Gene ID
100328839
Gene Name
olfactory receptor, family 52, subfamily J, member 3
Gene Aliases
-
Chromosome Location
Chr.1: 145819495 - 145820433 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171440.1 NP_001164911.1 1 - 1 647 74

View Full Product Details