Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADH2-2
Entrez Gene ID
100345521
Gene Name
alcohol dehydrogenase class-2 isozyme 2
Gene Aliases
-
Chromosome Location
Chr.15: 50725509 - 50749287 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184829.1 NP_001171758.1 8 - 9 1170 100
GenBank mRNA AJ002389.1 - 8 - 9 1170 100

View Full Product Details