Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CALM2
Entrez Gene ID
100352392
Gene Name
calmodulin 2
Gene Aliases
CALM, CAM
Chromosome Location
Chr.2: 139151338 - 139167852 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195640.1 NP_001182569.1 2 - 3 191 76

View Full Product Details