Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB3
Entrez Gene ID
100340734
Gene Name
ankyrin repeat and SOCS box containing 3
Gene Aliases
-
Chromosome Location
Chr.2: 132448716 - 132550409 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205047.1 NP_001191976.1 5 - 6 803 72
XM_008252739.2 XP_008250961.1 5 - 6 855 72

View Full Product Details