Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP2C16
Entrez Gene ID
100328938
Gene Name
cytochrome P450 2C16
Gene Aliases
-
Chromosome Location
Chr.18: 43043973 - 43071634 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171122.1 NP_001164593.1 2 - 3 394 86
GenBank mRNA M29661.1 - 2 - 3 394 86
M29968.1 - 2 - 3 394 86

View Full Product Details