Gene Transcripts

Gene Symbol
Rnf112
Entrez Gene ID
24916
Gene Name
ring finger protein 112
Gene Aliases
BFP, Bfb, Zfp179, Znf179
Chromosome Location
Chr. 10: 47719034 - 47726950 on Build Rnor_6.0
UniGene ID
Rn.7544
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138613.1 NP_619516.1 2, 3 189
XM_006246441.2 XP_006246503.1 4, 5 1301
XM_006246445.2 XP_006246507.1 2, 3 192
GenBank mRNA AF054586.1 - 2, 3 189

View Full Product Details