Gene Transcripts

Gene Symbol
Adrbk2
Entrez Gene ID
25372
Gene Name
adrenergic, beta, receptor kinase 2
Gene Aliases
Grk3
Chromosome Location
Chr. 12: 49626871 - 49750389 on Build Rnor_6.0
UniGene ID
Rn.9861
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012897.2 NP_037029.1 4 283
XM_008769317.1 XP_008767539.1 3 174

View Full Product Details