Gene Transcripts

Gene Symbol
Hmbs
Entrez Gene ID
25709
Gene Name
hydroxymethylbilane synthase
Gene Aliases
PBG-D, PBGD, URO-S, hemC
Chromosome Location
Chr. 8: 48667278 - 48674670 on Build Rnor_6.0
UniGene ID
Rn.11080
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013168.2 NP_037300.2 11 811
XM_006242896.1 XP_006242958.1 11 897
XM_006242897.1 XP_006242959.1 10 819
XM_008766148.1 XP_008764370.1 10 981
XM_008766149.1 XP_008764371.1 10 918
GenBank mRNA BC088162.1 - 11 811
FQ225972.1 - 5 296
FQ226062.1 - 11 734
FQ230962.1 - 5 296
FQ234793.1 - 3 183
FQ234870.1 - 9 967
X06827.1 - 11 686
Y12006.1 - 11 681

View Full Product Details