Gene Transcripts

Gene Symbol
Pvrl3
Entrez Gene ID
288124
Gene Name
poliovirus receptor-related 3
Gene Aliases
Chromosome Location
Chr. 11: 57897879 - 57995192 on Build Rnor_6.0
UniGene ID
Rn.56373
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105883.1 NP_001099353.1 3 645
XM_006248335.2 XP_006248397.1 3 683
XM_006248337.1 XP_006248399.1 3 683
XM_006248338.2 XP_006248400.1 3 683
XM_008768723.1 XP_008766945.1 3 626
XM_008768724.1 XP_008766946.1 3 1340

View Full Product Details