Gene Transcripts

Gene Symbol
Afap1l1
Entrez Gene ID
291565
Gene Name
actin filament associated protein 1-like 1
Gene Aliases
RGD1311580
Chromosome Location
Chr. 18: 57052662 - 57114580 on Build Rnor_6.0
UniGene ID
Rn.224736
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106142.1 NP_001099612.1 7 751
XM_006254794.2 XP_006254856.1 7 786
GenBank mRNA FQ211638.1 - 7 716
FQ231300.1 - 769

View Full Product Details