Gene Transcripts

Gene Symbol
Pabpc6
Entrez Gene ID
292295
Gene Name
poly(A) binding protein, cytoplasmic 6
Gene Aliases
Pabpc3
Chromosome Location
Chr. 1: 51619875 - 51622951 on Build Rnor_6.0
UniGene ID
Rn.108830
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106208.2 NP_001099678.1 1 566
GenBank mRNA BC168959.1 - 1 566

View Full Product Details