Gene Transcripts

Gene Symbol
Chsy1
Entrez Gene ID
292999
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
Chromosome Location
Chr. 1: 127010587 - 127071570 on Build Rnor_6.0
UniGene ID
Rn.13788
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106268.1 NP_001099738.1 3 2069

View Full Product Details