Gene Transcripts

Gene Symbol
Art2b
Entrez Gene ID
293152
Gene Name
ADP-ribosyltransferase 2b
Gene Aliases
ARTC2, Ag-F, Art2, Art2a, LY2, Ly-2, PtaA, RT6.1, RT6.2, Rt6
Chromosome Location
Chr. 1: 166667304 - 166685081 on Build Rnor_6.0
UniGene ID
Rn.107075
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198735.2 NP_942030.1 2 415
XM_008759694.1 XP_008757916.1 2 643
XM_008759695.1 XP_008757917.1 2 643
XM_008759696.1 XP_008757918.1 2 1264
XM_008759697.1 XP_008757919.1 3 1632
XM_008759698.1 XP_008757920.1 5 1004
XM_008759699.1 XP_008757921.1 2 490
XM_008759700.1 XP_008757922.1 4 735
XM_008759701.1 XP_008757923.1 2 558
XM_008759702.1 XP_008757924.1 2 1264
GenBank mRNA BC099070.1 - 2 415
FQ227811.1 - 1 188
FQ230306.1 - 4 751
M85193.1 - 2 392
X52082.1 - 2 385
X99123.1 - 3 465

View Full Product Details