Gene Transcripts

Gene Symbol
Olr70
Entrez Gene ID
293215
Gene Name
olfactory receptor 70
Gene Aliases
Chromosome Location
Chr. 1: 168159870 - 168160811 on Build Rnor_6.0
UniGene ID
Rn.141913
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001269.1 NP_001001269.1 1 823

View Full Product Details