Gene Transcripts

Gene Symbol
Olr80
Entrez Gene ID
293223
Gene Name
olfactory receptor 80
Gene Aliases
Chromosome Location
Chr. 1: 168282806 - 168283759 on Build Rnor_6.0
UniGene ID
Rn.141906
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001270.1 NP_001001270.1 1 514

View Full Product Details