Gene Transcripts

Gene Symbol
tGap1
Entrez Gene ID
294892
Gene Name
GTPase activating protein testicular GAP1
Gene Aliases
Chromosome Location
Chr. 2: 92552805 - 92625921 on Build Rnor_6.0
UniGene ID
Rn.162562
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007635.2 NP_001007636.2 13 1724
XM_006232134.2 XP_006232196.1 15 2066
GenBank mRNA AY631396.1 - 13 1724
AY631398.1 - 6 868
AY631399.1 - 13 1489

View Full Product Details