Gene Transcripts

Gene Symbol
Spata16
Entrez Gene ID
294932
Gene Name
spermatogenesis associated 16
Gene Aliases
RGD1563726
Chromosome Location
Chr. 2: 112281328 - 112659911 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760900.1 XP_008759122.1 7, 8 1397
XM_008760901.1 XP_008759123.1 7, 8 1360

View Full Product Details