Gene Transcripts

Gene Symbol
Baat
Entrez Gene ID
29725
Gene Name
bile acid CoA:amino acid N-acyltransferase
Gene Aliases
kan-1
Chromosome Location
Chr. 5: 64768397 - 64777368 on Build Rnor_6.0
UniGene ID
Rn.11129
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017300.2 NP_058996.2 2 222
GenBank mRNA BC088153.1 - 2 222
D43964.1 - 2 237
FQ218773.1 - 266

View Full Product Details