Gene Transcripts

Gene Symbol
Sbf1
Entrez Gene ID
300147
Gene Name
SET binding factor 1
Gene Aliases
Chromosome Location
Chr. 7: 130261552 - 130288562 on Build Rnor_6.0
UniGene ID
Rn.7632
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271173.1 NP_001258102.1 25 3330
XM_006242184.2 XP_006242246.1 25 3383
XM_006242185.2 XP_006242247.1 23 2990

View Full Product Details