Gene Transcripts

Gene Symbol
Tfap2b
Entrez Gene ID
301285
Gene Name
transcription factor AP-2 beta
Gene Aliases
Tcfap2b
Chromosome Location
Chr. 9: 25410669 - 25440426 on Build Rnor_6.0
UniGene ID
Rn.12263
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106896.2 NP_001100366.2 4 868
XM_006244649.1 XP_006244711.1 4 868
XM_008766927.1 XP_008765149.1 5 847

View Full Product Details