Gene Transcripts

Gene Symbol
Mageb3
Entrez Gene ID
302746
Gene Name
melanoma antigen family B, 3
Gene Aliases
RGD1562343
Chromosome Location
Chr. X: 54843202 - 54844197 on Build Rnor_6.0
UniGene ID
Rn.218617
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106965.1 NP_001100435.1 1 733

View Full Product Details