Gene Transcripts

Gene Symbol
Kif19
Entrez Gene ID
303659
Gene Name
kinesin family member 19
Gene Aliases
Kif19a, RGD1559936
Chromosome Location
Chr. 10: 103308392 - 103334466 on Build Rnor_6.0
UniGene ID
Rn.101489
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271202.1 NP_001258131.1 6 559

View Full Product Details