Gene Transcripts

Gene Symbol
Urb1
Entrez Gene ID
304104
Gene Name
URB1 ribosome biogenesis 1 homolog (S. cerevisiae)
Gene Aliases
RGD1304621
Chromosome Location
Chr. 11: 30916740 - 30977423 on Build Rnor_6.0
UniGene ID
Rn.95642
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191661.1 NP_001178590.1 8 921

View Full Product Details