Gene Transcripts

Gene Symbol
Tp53bp2
Entrez Gene ID
305025
Gene Name
tumor protein p53 binding protein, 2
Gene Aliases
Trp53bp2
Chromosome Location
Chr. 13: 100817204 - 100873837 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_223012.8 XP_223012.6 16 3231

View Full Product Details