Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013130.1 | NP_001013148.1 | 10 | 1242 | |
XM_008770458.1 | XP_008768680.1 | 10 | 2779 | ||
XM_008770460.1 | XP_008768682.1 | 9 | 2636 | ||
XM_008770461.1 | XP_008768683.1 | 10 | 1259 | ||
XM_008770462.1 | XP_008768684.1 | 9 | 2636 | ||
XM_008770463.1 | XP_008768685.1 | 10 | 2779 | ||
GenBank mRNA | AY238344.1 | - | 10 | 1242 |