Gene Transcripts

Gene Symbol
Slitrk5
Entrez Gene ID
306152
Gene Name
SLIT and NTRK-like family, member 5
Gene Aliases
Chromosome Location
Chr. 15: 96817892 - 96826760 on Build Rnor_6.0
UniGene ID
Rn.46737
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107284.1 NP_001100754.1 1 1939
XM_006252444.2 XP_006252506.1 2 2544
XM_006252445.2 XP_006252507.1 2 2207
XM_006252446.2 XP_006252508.1 2 2017
XM_006252447.2 XP_006252509.1 2 2313

View Full Product Details