Gene Transcripts

Gene Symbol
Hiatl1
Entrez Gene ID
306687
Gene Name
hippocampus abundant transcript-like 1
Gene Aliases
RGD1308377
Chromosome Location
Chr. 17: 2607777 - 2660902 on Build Rnor_6.0
UniGene ID
Rn.204848
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107334.1 NP_001100804.1 5 854
GenBank mRNA BC166784.1 - 5 847
FQ230882.1 - 5 859

View Full Product Details