Gene Transcripts

Gene Symbol
Dsp
Entrez Gene ID
306871
Gene Name
desmoplakin
Gene Aliases
Chromosome Location
Chr. 17: 27286811 - 27334451 on Build Rnor_6.0
UniGene ID
Rn.231913
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_225259.8 XP_225259.4 24 8155
GenBank mRNA BC098071.1 - 1 1086

View Full Product Details