Gene Transcripts

Gene Symbol
Prl7a4
Entrez Gene ID
306929
Gene Name
prolactin family 7, subfamily a, member 4
Gene Aliases
Chromosome Location
Chr. 17: 39095463 - 39104519 on Build Rnor_6.0
UniGene ID
Rn.31448
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008341.1 NP_001008342.1 5, 6 584
GenBank mRNA AY741310.1 - 5, 6 584

View Full Product Details