Gene Transcripts

Gene Symbol
Cmtm2a
Entrez Gene ID
307616
Gene Name
CKLF-like MARVEL transmembrane domain containing 2A
Gene Aliases
Cklfsf1
Chromosome Location
Chr. 19: 880024 - 887144 on Build Rnor_6.0
UniGene ID
Rn.163394
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013142.1 NP_001013160.1 4 551
GenBank mRNA BC078975.1 - 4 551

View Full Product Details