Gene Transcripts

Gene Symbol
Trim67
Entrez Gene ID
307938
Gene Name
tripartite motif-containing 67
Gene Aliases
RGD1560667
Chromosome Location
Chr. 19: 57556403 - 57592722 on Build Rnor_6.0
UniGene ID
Rn.221884
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135715.1 NP_001129187.1 8 2278
XM_006255815.2 XP_006255877.1 8 2569

View Full Product Details