Gene Transcripts

Gene Symbol
Ifi44l
Entrez Gene ID
310968
Gene Name
interferon-induced protein 44-like
Gene Aliases
H28
Chromosome Location
Chr. 2: 256948722 - 256979573 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_227820.8 XP_227820.6 6 1150

View Full Product Details