Gene Transcripts

Gene Symbol
Pax1
Entrez Gene ID
311505
Gene Name
paired box 1
Gene Aliases
Pax-1
Chromosome Location
Chr. 3: 141577504 - 141589849 on Build Rnor_6.0
UniGene ID
Rn.40296
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107787.1 NP_001101257.1 3 1363
XM_006235152.2 XP_006235214.2 4 2669

View Full Product Details