Gene Transcripts

Gene Symbol
Cyp2j10
Entrez Gene ID
313373
Gene Name
cytochrome P450, family 2, subfamily j, polypeptide 10
Gene Aliases
RGD1563215
Chromosome Location
Chr. 5: 115067847 - 115101961 on Build Rnor_6.0
UniGene ID
Rn.34638
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134980.1 NP_001128452.1 6 1086
GenBank mRNA BC168751.1 - 6 1086

View Full Product Details