Gene Transcripts

Gene Symbol
Rgp1
Entrez Gene ID
313493
Gene Name
RGP1 retrograde golgi transport homolog (S. cerevisiae)
Gene Aliases
RGD1311757
Chromosome Location
Chr. 5: 59080380 - 59088777 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238138.2 XP_006238200.1 3 294

View Full Product Details