Gene Transcripts

Gene Symbol
Nt5c1a
Entrez Gene ID
313574
Gene Name
5'-nucleotidase, cytosolic IA
Gene Aliases
Chromosome Location
Chr. 5: 140888986 - 140904009 on Build Rnor_6.0
UniGene ID
Rn.105265
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107976.1 NP_001101446.1 3 411

View Full Product Details