Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
314288
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
Chromosome Location
Chr. 6: 105611430 - 105747176 on Build Rnor_6.0
UniGene ID
Rn.23381
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170347.1 NP_001163818.1 33 6300
XM_006240299.2 XP_006240361.1 33 6311
XM_006240300.2 XP_006240362.1 33 6308
XM_006240301.1 XP_006240363.1 34 6347
XM_006240302.2 XP_006240364.1 31 5993
XM_008764788.1 XP_008763010.1 32 6221
XM_008764789.1 XP_008763011.1 33 7707

View Full Product Details