Gene Transcripts

Gene Symbol
Flvcr2
Entrez Gene ID
314323
Gene Name
feline leukemia virus subgroup C cellular receptor family, member 2
Gene Aliases
Chromosome Location
Chr. 6: 109617348 - 109681495 on Build Rnor_6.0
UniGene ID
Rn.92663
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199109.2 NP_954540.2 8 1724
GenBank mRNA AY260573.1 - 8 1476
BC101899.1 - 8 1724 7932187

View Full Product Details