Gene Transcripts

Gene Symbol
Asap1
Entrez Gene ID
314961
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 1
Gene Aliases
Ddef1
Chromosome Location
Chr. 7: 104670076 - 104951090 on Build Rnor_6.0
UniGene ID
Rn.63466
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044245.1 NP_001037710.1 10 1000
GenBank mRNA DQ238622.1 - 10 1000
DQ238623.1 - 9 1000
DQ238624.1 - 9 964

View Full Product Details